Assay Details
Gene Symbol: Gria4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113180.1 | NP_001106651.1 | 5 - 6 | 1155 | 123 | |
NM_001113181.1 | NP_001106652.1 | 5 - 6 | 1155 | 123 | ||
NM_019691.4 | NP_062665.3 | 5 - 6 | 1155 | 123 | ||
GenBank mRNA | AB022913.1 | - | 5 - 6 | 889 | 123 | |
AB079073.1 | - | 4 - 5 | 676 | 123 | ||
AB102777.1 | - | 4 - 5 | 676 | 123 | ||
AK031568.1 | - | 5 - 6 | 1129 | 123 | ||
AK032103.1 | - | 5 - 6 | 1156 | 123 | ||
AK135034.1 | - | 5 - 6 | 861 | 123 | ||
BC060697.1 | - | 5 - 6 | 1063 | 6409918 | 123 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
