Assay Details
Gene Symbol: Neurl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082974.2 | NP_001076443.1 | 1 - 2 | 903 | 85 | |
GenBank mRNA | AK004524.1 | - | 1 - 2 | 764 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P418935 | NM_001082974.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P418935 | AK004524.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P418935 | NM_001082974.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P418935 | AK004524.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 422037 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P418935 | NM_001082974.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P418935 | AK004524.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P418935 | NM_001082974.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P418935 | AK004524.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P00550503 | - | - |
Whole Mouse Genome G4122A | A_51_P418935 | NM_001082974.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P418935 | AK004524.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P418935 | NM_001082974.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P418935 | AK004524.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P108327 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01725833 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01725858 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01725853 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01725854 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P01725845 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820028 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820035 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820048 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820031 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819952 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820033 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820023 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820063 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819984 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820046 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820040 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820039 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819999 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820030 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820044 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820054 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820021 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819985 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820032 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820056 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820050 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21819950 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820001 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P21820026 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820042 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820035 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01725850 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01682189 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820066 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820068 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02201770 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820001 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820041 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01947791 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820028 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820063 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02108227 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02201774 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01725855 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820023 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820046 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01682185 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820059 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01740501 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820051 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01725854 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820026 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02201772 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P21820033 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P01682190 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02108225 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02265151 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02108226 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02281951 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10489568 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10489567 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10478596 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10478595 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1429141_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10489566 | - | - |
Mouse Genome 430 2.0 Array | 1429141_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1429141_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 415115_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 415115_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0200043770.mm.1 | TC0200005135.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200043770.mm.1 | NM_001082974.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200043770.mm.1 | NM_001082974.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200043771.mm.1 | TC0200005135.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200043771.mm.1 | NM_001082974.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200043771.mm.1 | NM_001082974.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200043773.mm.1 | TC0200005135.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0200043773.mm.1 | NM_001082974.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0200043773.mm.1 | NM_001082974.2 | Refseq |
Mouse Expression Set 430 | 1429141_at | - | - |
Mouse Expression Set 430 | 1429141_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429141_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1429141_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 442176 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.0 expression beadchip | 102370093 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1256950 | - | - |
Illumina Sentrix mouse-6.v1 bead array | 102370093 | - | - |
Illumina Sentrix Mouse-6 Expression BeadChip (2) | 102370093 | - | - |
Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1256950 | - | - |
