Assay Details
Gene Symbol: Ndrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013865.2 | NP_038893.1 | 4 - 5 | 352 | 96 | |
NM_180956.1 | NP_851287.1 | 4 - 5 | 352 | 96 | ||
GenBank mRNA | AB033922.1 | - | 4 - 5 | 266 | 96 | |
AK002272.1 | - | 3 - 4 | 266 | 96 | ||
AK004689.1 | - | 4 - 5 | 298 | 96 | ||
AK036277.1 | - | 4 - 5 | 277 | 96 | ||
AK037487.1 | - | 4 - 5 | 278 | 96 | ||
AK154927.1 | - | 4 - 5 | 319 | 96 | ||
AK209466.1 | - | 1 - 2 | 66 | 96 | ||
BC018504.1 | - | 4 - 5 | 286 | 4500970 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
