Assay Details
Gene Symbol: Ndrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013865.2 | NP_038893.1 | 9 - 10 | 741 | 81 | |
NM_180956.1 | NP_851287.1 | 9 - 10 | 741 | 81 | ||
GenBank mRNA | AB033922.1 | - | 9 - 10 | 655 | 81 | |
AK002272.1 | - | 8 - 9 | 655 | 81 | ||
AK004689.1 | - | 9 - 10 | 687 | 81 | ||
AK036277.1 | - | 9 - 10 | 666 | 81 | ||
AK037487.1 | - | 9 - 10 | 667 | 81 | ||
AK154927.1 | - | 9 - 10 | 708 | 81 | ||
AK214669.1 | - | 5 - 6 | 81 | |||
AK218281.1 | - | 5 - 6 | 311 | 81 | ||
BC018504.1 | - | 9 - 10 | 675 | 4500970 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
