Assay Details
Gene Symbol: Ndrg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013865.2 | NP_038893.1 | 10 - 11 | 853 | 96 | |
NM_180956.1 | NP_851287.1 | 10 - 11 | 853 | 96 | ||
GenBank mRNA | AB033922.1 | - | 10 - 11 | 767 | 96 | |
AK002272.1 | - | 9 - 10 | 767 | 96 | ||
AK004689.1 | - | 10 - 11 | 799 | 96 | ||
AK036277.1 | - | 10 - 11 | 778 | 96 | ||
AK037487.1 | - | 10 - 11 | 779 | 96 | ||
AK154927.1 | - | 10 - 11 | 820 | 96 | ||
BC018504.1 | - | 10 - 11 | 787 | 4500970 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
