Assay Details
Gene Symbol: Man2b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010764.2 | NP_034894.2 | 4 - 5 | 688 | 65 | |
GenBank mRNA | AK004817.2 | - | 4 - 5 | 645 | 65 | |
AK035551.1 | - | 4 - 5 | 643 | 65 | ||
AK088826.1 | - | 4 - 5 | 645 | 65 | ||
AK147928.1 | - | 4 - 5 | 646 | 65 | ||
AK171166.1 | - | 4 - 5 | 646 | 65 | ||
AK196301.1 | - | 2 - 3 | 274 | 65 | ||
BC005430.1 | - | 4 - 5 | 633 | 3488681 | 65 | |
U29947.1 | - | 4 - 5 | 631 | 65 | ||
U87240.1 | - | 4 - 5 | 622 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
