Assay Details
Gene Symbol: Pecam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK037551.1 | - | 8 - 9 | 2073 | 71 | |
AK084527.1 | - | 7 - 8 | 1663 | 71 | ||
AK169431.1 | - | 6 - 7 | 1362 | 71 | ||
BC008519.1 | - | 7 - 8 | 1667 | 3592726 | 71 | |
BC085502.1 | - | 7 - 8 | 1469 | 5720681 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
