Assay Details
Gene Symbol: Parpbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029249.2 | NP_083525.1 | 6 - 7 | 1267 | 127 | |
GenBank mRNA | AK030970.1 | - | 1 - 2 | 96 | 127 | |
AK131981.1 | - | 6 - 7 | 1253 | 127 | ||
AK137160.1 | - | 6 - 7 | 1216 | 127 | ||
AK150571.1 | - | 6 - 7 | 1231 | 127 | ||
AK151268.1 | - | 6 - 7 | 1231 | 127 | ||
AK152208.1 | - | 6 - 7 | 1231 | 127 | ||
BC050991.1 | - | 6 - 7 | 1211 | 127 | ||
BC070476.1 | - | 6 - 7 | 1267 | 30650782 | 127 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK030970.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
