Assay Details
Gene Symbol: Ap1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301211.1 | NP_001288140.1 | 10 - 11 | 1358 | 129 | |
NM_009677.6 | NP_033807.2 | 11 - 12 | 1367 | 129 | ||
GenBank mRNA | AK034198.1 | - | 8 - 9 | 596 | 129 | |
AK036387.1 | - | 11 - 12 | 1310 | 129 | ||
AK145821.1 | - | 10 - 11 | 1289 | 129 | ||
BC006691.1 | - | 10 - 11 | 1034 | 129 | ||
BC052703.1 | - | 11 - 12 | 1240 | 5698075 | 129 | |
BC054535.1 | - | 11 - 12 | 1240 | 5698075 | 129 | |
X54424.1 | - | 10 - 11 | 1280 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
