Assay Details
Gene Symbol: Ap1g1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301211.1 | NP_001288140.1 | 7 - 8 | 1124 | 126 | |
NM_009677.6 | NP_033807.2 | 8 - 9 | 1133 | 126 | ||
GenBank mRNA | AK034198.1 | - | 5 - 6 | 362 | 126 | |
AK036387.1 | - | 8 - 9 | 1076 | 126 | ||
AK145821.1 | - | 7 - 8 | 1055 | 126 | ||
BC006691.1 | - | 7 - 8 | 800 | 126 | ||
BC052703.1 | - | 8 - 9 | 1006 | 5698075 | 126 | |
BC054535.1 | - | 8 - 9 | 1006 | 5698075 | 126 | |
X54424.1 | - | 7 - 8 | 1046 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
