Assay Details
Gene Symbol: Mcm5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK033196.1 | - | 10 - 11 | 1414 | 88 | |
AK088558.1 | - | 3 - 4 | 330 | 88 | ||
AK133826.1 | - | 10 - 11 | 1415 | 88 | ||
AK214383.1 | - | 2 - 3 | 270 | 88 | ||
BC090645.1 | - | 10 - 11 | 1367 | 88 | ||
BC094416.1 | - | 10 - 11 | 1379 | 5707724 | 88 | |
D26090.1 | - | 9 - 10 | 1434 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
