Assay Details
Gene Symbol: Rbck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019705.3 | NP_062679.2 | 1 - 2 | 142 | 65 | |
GenBank mRNA | BC034555.1 | - | 1 - 2 | 138 | 4192310 | 65 |
BY267597.1 | - | 1 - 2 | 121 | 65 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC034555.1, NM_019705.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
