Assay Details
Gene Symbol: Nup88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083331.2 | NP_001076800.1 | 14 - 15 | 1969 | 82 | |
NM_001276406.1 | NP_001263335.1 | 14 - 15 | 1951 | 82 | ||
NM_172394.3 | NP_765982.2 | 14 - 15 | 1936 | 82 | ||
NR_075087.1 | 13 - 14 | 2395 | 82 | |||
GenBank mRNA | AJ532593.1 | - | 14 - 15 | 1945 | 82 | |
AK002643.1 | - | 14 - 15 | 1937 | 82 | ||
AK028563.1 | - | 14 - 15 | 1959 | 82 | ||
AK050893.1 | - | 11 - 12 | 2373 | 82 | ||
AK085776.1 | - | 14 - 15 | 1935 | 82 | ||
AK139738.1 | - | 13 - 14 | 2303 | 82 | ||
AK160619.1 | - | 14 - 15 | 1937 | 82 | ||
AK166214.1 | - | 14 - 15 | 1941 | 82 | ||
BC032929.1 | - | 14 - 15 | 1917 | 1379673 | 82 | |
U01135.1 | - | 2 - 3 | 158 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
