Assay Details
Gene Symbol: C1qtnf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204129.1 | NP_001191058.1 | 1 - 2 | 108 | 85 | |
GenBank mRNA | AK005484.1 | - | 1 - 2 | 67 | 85 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK005484.1, NM_001204129.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
