Assay Details
Gene Symbol: Kansl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081045.1 | NP_001074514.1 | 11 - 12 | 3165 | 84 | |
GenBank mRNA | AK006970.1 | - | 3 - 4 | 276 | 84 | |
AK122484.1 | - | 11 - 12 | 3171 | 84 | ||
BC043121.1 | - | 11 - 12 | 2714 | 6409084 | 84 | |
BC054752.1 | - | 12 - 13 | 2988 | 6834541 | 84 | |
BC079594.1 | - | 11 - 12 | 2713 | 6827379 | 84 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
