Assay Details
Gene Symbol: Psmb10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013640.3 | NP_038668.2 | 6 - 7 | 945 | 58 | |
GenBank mRNA | BC004730.1 | - | 6 - 7 | 671 | 3583052 | 58 |
CT010274.1 | - | 6 - 7 | 563 | 58 | ||
D85561.1 | - | 6 - 7 | 640 | 58 | ||
U77784.1 | - | 6 - 7 | 632 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
