Assay Details
Gene Symbol: Trmt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164559.1 | NP_001158031.1 | 2 - 3 | 380 | 63 | |
NM_001164560.1 | NP_001158032.1 | 2 - 3 | 304 | 63 | ||
NM_198020.2 | NP_932137.2 | 2 - 3 | 407 | 63 | ||
GenBank mRNA | AK032945.1 | - | 2 - 3 | 315 | 63 | |
AK040715.1 | - | 2 - 3 | 270 | 63 | ||
AK078395.1 | - | 2 - 3 | 345 | 63 | ||
AK084775.1 | - | 2 - 3 | 345 | 63 | ||
AK139463.1 | - | 2 - 3 | 339 | 63 | ||
AK155251.1 | - | 2 - 3 | 372 | 63 | ||
AK159467.1 | - | 2 - 3 | 345 | 63 | ||
AK170412.1 | - | 2 - 3 | 269 | 63 | ||
AK171002.1 | - | 2 - 3 | 372 | 63 | ||
BC027404.1 | - | 2 - 3 | 376 | 4950980 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
