Assay Details
Gene Symbol: Nfat5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286260.1 | NP_001273189.1 | 11 - 12 | 2215 | 127 | |
NM_018823.2 | NP_061293.2 | 12 - 13 | 2269 | 127 | ||
NM_133957.3 | NP_598718.2 | 13 - 14 | 2331 | 127 | ||
GenBank mRNA | AF162853.1 | - | 11 - 12 | 2126 | 127 | |
AF200687.1 | - | 12 - 13 | 1904 | 127 | ||
AF228706.1 | - | 9 - 10 | 1969 | 127 | ||
AF369980.1 | - | 13 - 14 | 1970 | 127 | ||
AF453571.2 | - | 13 - 14 | 1939 | 127 | ||
AK122381.1 | - | 3 - 4 | 356 | 127 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
