Assay Details
Gene Symbol: Sulf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252578.1 | NP_001239507.1 | 18 - 19 | 2940 | 136 | |
NM_001252579.1 | NP_001239508.1 | 18 - 19 | 2786 | 136 | ||
NM_028072.5 | NP_082348.2 | 18 - 19 | 2782 | 136 | ||
GenBank mRNA | AK008108.1 | - | 9 - 10 | 1045 | 136 | |
AK028874.1 | - | 18 - 19 | 2697 | 136 | ||
AK034712.1 | - | 18 - 19 | 2767 | 136 | ||
AK036685.1 | - | 18 - 19 | 2718 | 136 | ||
AK049170.1 | - | 13 - 14 | 1697 | 136 | ||
AK081643.1 | - | 18 - 19 | 2740 | 136 | ||
AK129316.1 | - | 18 - 19 | 2940 | 136 | ||
AK133336.1 | - | 2756 | 136 | |||
AK165183.1 | - | 18 - 19 | 2694 | 136 | ||
AY101177.1 | - | 17 - 18 | 2601 | 136 | ||
BC027238.1 | - | 7 - 8 | 756 | 136 | ||
BC062900.1 | - | 18 - 19 | 2738 | 136 | ||
BC141086.1 | - | 17 - 18 | 2535 | 9055698 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
