Assay Details
Gene Symbol: Sulf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252578.1 | NP_001239507.1 | 9 - 10 | 1685 | 76 | |
NM_001252579.1 | NP_001239508.1 | 9 - 10 | 1531 | 76 | ||
NM_028072.5 | NP_082348.2 | 9 - 10 | 1527 | 76 | ||
GenBank mRNA | AK028874.1 | - | 9 - 10 | 1442 | 76 | |
AK034712.1 | - | 9 - 10 | 1512 | 76 | ||
AK036685.1 | - | 9 - 10 | 1463 | 76 | ||
AK049170.1 | - | 4 - 5 | 442 | 76 | ||
AK081643.1 | - | 9 - 10 | 1485 | 76 | ||
AK129316.1 | - | 9 - 10 | 1685 | 76 | ||
AK133336.1 | - | 1501 | 76 | |||
AK165183.1 | - | 9 - 10 | 1439 | 76 | ||
AY101177.1 | - | 8 - 9 | 1346 | 76 | ||
BC062900.1 | - | 9 - 10 | 1484 | 76 | ||
BC141086.1 | - | 8 - 9 | 1280 | 9055698 | 76 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
