Assay Details
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242558.1 | NP_001229487.1 | 10 - 11 | 3141 | 89 | |
NM_019825.3 | NP_062799.2 | 11 - 12 | 6117 | 89 | ||
GenBank mRNA | AF216186.1 | - | 10 - 11 | 6002 | 89 | |
AK129079.1 | - | 4 - 5 | 3276 | 89 | ||
AK136585.1 | - | 1 - 2 | 889 | 89 | ||
AK142566.1 | - | 5 - 6 | 4302 | 89 | ||
BC083187.1 | - | 11 - 12 | 6117 | 30544739 | 89 | |
JF707634.1 | - | 9 - 10 | 3000 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
