Assay Details
Gene Symbol: Ncoa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242558.1 | NP_001229487.1 | 2 - 3 | 377 | 92 | |
NM_019825.3 | NP_062799.2 | 2 - 3 | 377 | 92 | ||
GenBank mRNA | AF216186.1 | - | 1 - 2 | 268 | 92 | |
AK144338.1 | - | 4 - 5 | 616 | 92 | ||
AK171864.1 | - | 2 - 3 | 538 | 92 | ||
BC031113.1 | - | 2 - 3 | 381 | 92 | ||
BC083187.1 | - | 2 - 3 | 377 | 30544739 | 92 | |
JF707634.1 | - | 1 - 2 | 236 | 92 | ||
JF707635.1 | - | 1 - 2 | 236 | 92 | ||
JF707636.1 | - | 1 - 2 | 236 | 92 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
