Assay Details
Gene Symbol: Pld2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK143930.1 | - | 4 - 5 | 720 | 132 | |
AK170701.1 | - | 4 - 5 | 712 | 132 | ||
BC047268.1 | - | 4 - 5 | 644 | 132 | ||
BC068317.1 | - | 4 - 5 | 657 | 30094606 | 132 | |
U87557.1 | - | 4 - 5 | 522 | 132 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
