Assay Details
Gene Symbol: Mpp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007863.2 | NP_031889.2 | 10 - 11 | 1073 | 67 | |
GenBank mRNA | AF079366.1 | - | 10 - 11 | 1024 | 67 | |
AK032270.1 | - | 9 - 10 | 2347 | 67 | ||
AY236514.1 | - | 9 - 10 | 907 | 67 | ||
BC052003.1 | - | 1320 | 67 | |||
BC062642.1 | - | 10 - 11 | 1041 | 5718318 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
