Assay Details
Gene Symbol: Dlg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109752.1 | NP_001103222.1 | 6 - 7 | 811 | 151 | |
NM_007864.3 | NP_031890.1 | 6 - 7 | 820 | 151 | ||
GenBank mRNA | BC014807.1 | - | 6 - 7 | 556 | 4501403 | 151 |
D50621.1 | - | 6 - 7 | 562 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
