Assay Details
Gene Symbol: Rad23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001297606.1 | NP_001284535.1 | 2 - 3 | 337 | 120 | |
NM_001297607.1 | NP_001284536.1 | 2 - 3 | 337 | 120 | ||
NM_009010.5 | NP_033036.2 | 2 - 3 | 337 | 120 | ||
GenBank mRNA | AK038300.1 | - | 2 - 3 | 314 | 120 | |
AK045224.1 | - | 2 - 3 | 327 | 120 | ||
AK161875.1 | - | 2 - 3 | 314 | 120 | ||
AK165467.1 | - | 2 - 3 | 338 | 120 | ||
BC003846.1 | - | 2 - 3 | 325 | 120 | ||
BC138684.1 | - | 2 - 3 | 297 | 8861705 | 120 | |
BC145372.1 | - | 2 - 3 | 297 | 9053930 | 120 | |
X92410.1 | - | 2 - 3 | 237 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
