Assay Details
Gene Symbol: Mybbp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016776.2 | NP_058056.2 | 18 - 19 | 2481 | 57 | |
GenBank mRNA | AK083908.1 | - | 18 - 19 | 2507 | 57 | |
AK143469.1 | - | 18 - 19 | 2506 | 57 | ||
AK143487.1 | - | 18 - 19 | 2514 | 57 | ||
AK149747.1 | - | 13 - 14 | 1790 | 57 | ||
AK154016.1 | - | 16 - 17 | 2700 | 57 | ||
AK155074.1 | - | 18 - 19 | 2492 | 57 | ||
AK193402.1 | - | 1 - 2 | 60 | 57 | ||
AK197626.1 | - | 2 - 3 | 287 | 57 | ||
BC004078.1 | - | 8 - 9 | 965 | 57 | ||
BC015070.1 | - | 18 - 19 | 2482 | 57 | ||
BC048858.1 | - | 18 - 19 | 2481 | 5363740 | 57 | |
BC052889.1 | - | 18 - 19 | 2502 | 30057169 | 57 | |
BC060167.1 | - | 17 - 18 | 2577 | 57 | ||
U63648.1 | - | 18 - 19 | 2490 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
