Assay Details
Gene Symbol: Hsp90b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011631.1 | NP_035761.1 | 14 - 15 | 2119 | 71 | |
GenBank mRNA | AK005469.1 | - | 1 - 2 | 47 | 71 | |
AK031918.1 | - | 10 - 11 | 1471 | 71 | ||
AK146885.1 | - | 14 - 15 | 2185 | 71 | ||
AK150811.1 | - | 14 - 15 | 2139 | 71 | ||
AK151413.1 | - | 14 - 15 | 2136 | 71 | ||
AK153550.1 | - | 14 - 15 | 2139 | 71 | ||
AK159974.1 | - | 14 - 15 | 2144 | 71 | ||
AK160093.1 | - | 14 - 15 | 2138 | 71 | ||
AK160827.1 | - | 1559 | 71 | |||
AK165975.1 | - | 14 - 15 | 2139 | 71 | ||
AK168952.1 | - | 14 - 15 | 2139 | 71 | ||
AK169090.1 | - | 14 - 15 | 2139 | 71 | ||
BC010445.1 | - | 14 - 15 | 2051 | 3587178 | 71 | |
BC011439.1 | - | 14 - 15 | 2125 | 3707817 | 71 | |
J03297.1 | - | 14 - 15 | 2119 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
