Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400007439.mm.1 |
TC0400001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007439.mm.1 |
NM_001162980.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007439.mm.1 |
NM_001162980.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007439.mm.1 |
NM_001285952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007439.mm.1 |
NM_001285953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007439.mm.1 |
NR_104376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007438.mm.1 |
TC0400001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007438.mm.1 |
NM_001162980.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007438.mm.1 |
NM_001162980.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007438.mm.1 |
NM_001285952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007438.mm.1 |
NM_001285953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007438.mm.1 |
NR_104376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007449.mm.1 |
TC0400001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007449.mm.1 |
NM_001162980.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007449.mm.1 |
NM_001162980.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007449.mm.1 |
NM_001285952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007449.mm.1 |
NM_001285953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007449.mm.1 |
NR_104376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007446.mm.1 |
TC0400001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007446.mm.1 |
NM_001162980.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007446.mm.1 |
NM_001162980.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007446.mm.1 |
NM_001285952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007446.mm.1 |
NM_001285953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007446.mm.1 |
NR_104376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007447.mm.1 |
TC0400001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007447.mm.1 |
NM_001162980.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007447.mm.1 |
NM_001162980.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007447.mm.1 |
NM_001285952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007447.mm.1 |
NM_001285953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007447.mm.1 |
NR_104376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007440.mm.1 |
TC0400001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007440.mm.1 |
NM_001162980.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007440.mm.1 |
NM_001162980.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007440.mm.1 |
NM_001285952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007440.mm.1 |
NR_104376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007444.mm.1 |
TC0400001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007444.mm.1 |
NM_001285952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007448.mm.1 |
TC0400001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007448.mm.1 |
NM_001162980.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007448.mm.1 |
NM_001162980.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007448.mm.1 |
NM_001285952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007448.mm.1 |
NM_001285953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007448.mm.1 |
NR_104376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007441.mm.1 |
TC0400001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007441.mm.1 |
NM_001162980.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007441.mm.1 |
NM_001162980.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007441.mm.1 |
NM_001285952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007441.mm.1 |
NM_001285953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007441.mm.1 |
NR_104376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007442.mm.1 |
TC0400001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007442.mm.1 |
NM_001162980.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007442.mm.1 |
NM_001162980.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007442.mm.1 |
NM_001285952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007442.mm.1 |
NM_001285953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007442.mm.1 |
NR_104376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007445.mm.1 |
TC0400001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007445.mm.1 |
NM_001162980.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007445.mm.1 |
NM_001162980.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007445.mm.1 |
NM_001285952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007445.mm.1 |
NM_001285953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007445.mm.1 |
NR_104376.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007443.mm.1 |
TC0400001090.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400007443.mm.1 |
NM_001162980.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007443.mm.1 |
NM_001162980.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007443.mm.1 |
NM_001285952.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007443.mm.1 |
NM_001285953.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400007443.mm.1 |
NR_104376.1 |
Refseq |