Assay Details
Gene Symbol: 1700024P16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162980.1 | NP_001156452.1 | 9 - 10 | 1613 | 74 | |
NM_001285952.1 | NP_001272881.1 | 10 - 11 | 1691 | 74 | ||
NM_001285953.1 | NP_001272882.1 | 9 - 10 | 1546 | 74 | ||
NR_104376.1 | 9 - 10 | 1613 | 74 | |||
GenBank mRNA | AK028077.1 | - | 9 - 10 | 1553 | 74 | |
AK133762.1 | - | 9 - 10 | 1484 | 74 | ||
BC137723.1 | - | 9 - 10 | 1613 | 8860742 | 74 | |
BC144857.1 | - | 10 - 11 | 1691 | 9053405 | 74 | |
BC144858.1 | - | 9 - 10 | 1614 | 9053406 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
