Assay Details
Gene Symbol: Gtf3c3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033194.2 | NP_001028366.1 | 14 - 15 | 2111 | 125 | |
GenBank mRNA | AK145040.1 | - | 2115 | 125 | ||
AK160997.1 | - | 14 - 15 | 2099 | 125 | ||
AK165830.1 | - | 14 - 15 | 2112 | 125 | ||
BC141157.1 | - | 14 - 15 | 2101 | 9055769 | 125 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
