Assay Details
Gene Symbol: Ipo8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081113.1 | NP_001074582.1 | 21 - 22 | 2711 | 95 | |
GenBank mRNA | AK078030.1 | - | 3 - 4 | 384 | 95 | |
AK140566.1 | - | 3 - 4 | 1240 | 95 | ||
BC018530.1 | - | 1 - 2 | 132 | 95 | ||
BC024474.1 | - | 3 - 4 | 374 | 95 | ||
BC027360.1 | - | 4 - 5 | 533 | 95 | ||
BC034091.1 | - | 8 - 9 | 1003 | 95 | ||
BC151036.1 | - | 21 - 22 | 2579 | 9087949 | 95 | |
BC151052.1 | - | 21 - 22 | 2579 | 9087965 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
