Assay Details
Gene Symbol: Arnt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037737.2 | NP_001032826.1 | 16 - 17 | 1723 | 71 | |
NM_009709.4 | NP_033839.2 | 15 - 16 | 1678 | 71 | ||
GenBank mRNA | AK028546.1 | - | 15 - 16 | 1665 | 71 | |
AK040475.1 | - | 16 - 17 | 1725 | 71 | ||
AK049738.1 | - | 2 - 3 | 369 | 71 | ||
AK141362.1 | - | 16 - 17 | 1594 | 71 | ||
AK145415.1 | - | 15 - 16 | 1633 | 71 | ||
AK152473.1 | - | 16 - 17 | 1705 | 71 | ||
AK153355.1 | - | 16 - 17 | 1705 | 71 | ||
AK220458.1 | - | 12 - 13 | 1854 | 71 | ||
BC012870.1 | - | 16 - 17 | 1685 | 3964932 | 71 | |
U10325.1 | - | 16 - 17 | 1587 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
