Assay Details
Gene Symbol: Fgd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008001.4 | NP_032027.2 | 6 - 7 | 1846 | 80 | |
GenBank mRNA | AK148151.1 | - | 6 - 7 | 1847 | 80 | |
BC011462.1 | - | 6 - 7 | 1783 | 3674293 | 80 | |
U22325.1 | - | 6 - 7 | 1445 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
