Assay Details
Gene Symbol: Pde4dip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039376.2 | NP_001034465.2 | 15 - 16 | 1997 | 59 | |
NM_001289701.1 | NP_001276630.1 | 15 - 16 | 1997 | 59 | ||
NM_001289702.1 | NP_001276631.1 | 15 - 16 | 1997 | 59 | ||
NM_178080.4 | NP_835181.2 | 8 - 9 | 2505 | 59 | ||
NR_110360.1 | 8 - 9 | 3019 | 59 | |||
GenBank mRNA | AK028810.1 | - | 8 - 9 | 2472 | 59 | |
AK122286.1 | - | 8 - 9 | 2500 | 59 | ||
AK141917.1 | - | 15 - 16 | 2123 | 59 | ||
BC025653.1 | - | 8 - 9 | 2431 | 59 | ||
BC028274.1 | - | 15 - 16 | 2007 | 59 | ||
BC034266.1 | - | 8 - 9 | 1170 | 59 | ||
BC050783.1 | - | 8 - 9 | 2459 | 59 | ||
BC057350.1 | - | 8 - 9 | 2936 | 59 | ||
BC141172.1 | - | 8 - 9 | 2433 | 59 | ||
BC141185.1 | - | 15 - 16 | 1997 | 9055797 | 59 | |
BC145504.1 | - | 15 - 16 | 1997 | 9054067 | 59 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
