Assay Details
Gene Symbol: Tcerg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039474.1 | NP_001034563.1 | 2 - 3 | 323 | 129 | |
NM_001289526.1 | NP_001276455.1 | 2 - 3 | 323 | 129 | ||
NR_110344.1 | 2 - 3 | 323 | 129 | |||
GenBank mRNA | AB023485.1 | - | 2 - 3 | 287 | 129 | |
AK010571.1 | - | 2 - 3 | 306 | 129 | ||
AK013830.1 | - | 2 - 3 | 309 | 129 | ||
AK076993.1 | - | 2 - 3 | 301 | 129 | ||
AK150312.1 | - | 2 - 3 | 313 | 129 | ||
AK168434.1 | - | 2 - 3 | 301 | 129 | ||
BC016542.1 | - | 2 - 3 | 281 | 129 | ||
BC039185.1 | - | 2 - 3 | 282 | 3982736 | 129 | |
BC040284.1 | - | 2 - 3 | 286 | 4460736 | 129 | |
CJ052824.1 | - | 2 - 3 | 323 | 129 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
