Assay Details
Gene Symbol: Rere | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085492.1 | NP_001078961.1 | 7 - 8 | 1132 | 85 | |
GenBank mRNA | BC145182.1 | - | 6 - 7 | 902 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
