Assay Details
Gene Symbol: Lrmp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001281980.1 | NP_001268909.1 | 3 - 4 | 322 | 90 | |
NM_001281981.1 | NP_001268910.1 | 4 - 5 | 363 | 90 | ||
NM_008511.3 | NP_032537.3 | 3 - 4 | 594 | 90 | ||
GenBank mRNA | AK040180.1 | - | 3 - 4 | 595 | 90 | |
AK050646.1 | - | 3 - 4 | 1032 | 90 | ||
AK079932.1 | - | 3 - 4 | 552 | 90 | ||
AK153823.1 | - | 3 - 4 | 592 | 90 | ||
AK160009.1 | - | 3 - 4 | 554 | 90 | ||
AK169578.1 | - | 4 - 5 | 347 | 90 | ||
BC052909.1 | - | 3 - 4 | 530 | 30076553 | 90 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK050646.1, NM_001281981.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
