Assay Details
Gene Symbol: Fkbp15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045528.1 | NP_001038993.1 | 18 - 19 | 1927 | 82 | |
GenBank mRNA | AK049459.1 | - | 18 - 19 | 1872 | 82 | |
AK049509.1 | - | 18 - 19 | 1927 | 82 | ||
AK082565.1 | - | 18 - 19 | 1876 | 82 | ||
AK085087.1 | - | 18 - 19 | 1918 | 82 | ||
AK122343.1 | - | 11 - 12 | 1166 | 82 | ||
AK161705.1 | - | 18 - 19 | 1857 | 82 | ||
BC034098.1 | - | 8 - 9 | 864 | 82 | ||
BC060651.1 | - | 18 - 19 | 1841 | 82 | ||
BC137706.1 | - | 18 - 19 | 1861 | 8860725 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
