Assay Details
Gene Symbol: Ptprh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207270.2 | NP_997153.2 | 6 - 7 | 1104 | 77 | |
GenBank mRNA | AB217856.1 | - | 5 - 6 | 777 | 77 | |
AK078884.1 | - | 5 - 6 | 779 | 77 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AK078884.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
