Assay Details
Gene Symbol: Kcnab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252654.1 | NP_001239583.1 | 1 - 2 | 153 | 87 | |
NM_010598.3 | NP_034728.2 | 1 - 2 | 153 | 87 | ||
GenBank mRNA | AK143166.1 | - | 1 - 2 | 130 | 87 | |
BC039178.1 | - | 1 - 2 | 83 | 4503373 | 87 | |
U31908.1 | - | 1 - 2 | 90 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK143166.1, BC039178.1, NM_001252654.1, NM_010598.3, U31908.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
