Assay Details
Gene Symbol: Oacyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177028.3 | NP_796002.1 | 1 - 2 | 179 | 73 | |
GenBank mRNA | AK030604.1 | - | 1 - 2 | 179 | 73 | |
BC141265.1 | - | 1 - 2 | 176 | 9055877 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
