Assay Details
Gene Symbol: Oacyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177028.3 | NP_796002.1 | 4 - 5 | 531 | 89 | |
GenBank mRNA | AK030604.1 | - | 4 - 5 | 531 | 89 | |
AK030615.1 | - | 2 - 3 | 311 | 89 | ||
BC141265.1 | - | 4 - 5 | 528 | 9055877 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
