Assay Details
Gene Symbol: Ttc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172073.1 | NP_001165544.1 | 1 - 2 | 191 | 94 | |
GenBank mRNA | AK005923.1 | - | 1 - 2 | 192 | 94 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_001172073.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
