Assay Details
Gene Symbol: Gm19466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040349.1 | 1 - 2 | 218 | 62 | ||
GenBank mRNA | AK044363.1 | - | 1 - 2 | 218 | 62 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | centrosomal protein 120 | Cep120 |
A230075C01, AU016693, Ccdc100 |
Anti-sense | Chr. 18 - 53681723 - 53745603 on NCBI Build GRCm38 | |
Downstream |
casein kinase 1, gamma 3 |
Csnk1g3 |
3300002K07Rik, C330049O21Rik |
169.2kb | Sense | Chr. 18 - 53862082 - 53955684 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 396462 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 651846 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 137783 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 291627 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 384626 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469652 | AK030452.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469652 | AY227772.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469652 | BC089474.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469652 | BC100420.1 | genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 469652 | NM_177915.3 | refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 638920 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 713906 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 768314 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 503315 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 140356 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 285361 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649861 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 210739 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 15396 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 8623 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 123052 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09004071 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P03478360 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09004046 | - | - |
Whole Mouse Genome G4122A | A_52_P520883 | AK044363.1 | mouse_transcriptome |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714566 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13987999 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714711 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714710 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13785753 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714706 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714569 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13994301 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714571 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714606 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714613 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714650 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714577 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714596 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13938521 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714638 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714579 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714646 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714640 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714708 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13617253 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714576 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714636 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714622 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714601 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714628 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714581 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714591 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714556 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714599 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714616 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714568 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714627 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714618 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714575 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714702 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714614 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714589 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714605 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13804442 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714620 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714703 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13777534 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714583 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714641 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714704 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13815234 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714625 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714558 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714617 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714647 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714584 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714600 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714634 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714607 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14135762 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714562 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714632 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714587 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714588 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714642 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714649 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714635 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13815231 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714653 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714603 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714595 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13815236 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714586 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714712 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714594 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714563 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13640600 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714643 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714612 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13640699 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31714629 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13706142 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR1800003917.mm.1 | TC1800000541.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800003917.mm.1 | NR_040349.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800003917.mm.1 | NR_040349.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800003915.mm.1 | TC1800000541.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800003915.mm.1 | NR_040349.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800003915.mm.1 | NR_040349.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800003916.mm.1 | TC1800000541.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR1800003916.mm.1 | NR_040349.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800003916.mm.1 | NR_040349.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 558362 | - | - |
