Assay Details
Gene Symbol: C87499 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198663.3 | NP_941065.2 | 1 - 2 | 69 | 85 | |
GenBank mRNA | CA559642.1 | - | 1 - 2 | 74 | 85 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the NM_198663.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
