Assay Details
Gene Symbol: Sos2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135559.1 | NP_001129031.1 | 4 - 5 | 757 | 113 | |
GenBank mRNA | Z11664.1 | - | 3 - 4 | 401 | 113 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
