Assay Details
Gene Symbol: Inadl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005784.1 | NP_001005784.1 | 17 - 18 | 2253 | 88 | |
NM_172696.2 | NP_766284.2 | 17 - 18 | 2253 | 88 | ||
GenBank mRNA | AK077268.1 | - | 4 - 5 | 535 | 88 | |
AK147866.1 | - | 17 - 18 | 2202 | 88 | ||
AK153597.1 | - | 6 - 7 | 1108 | 88 | ||
AK156166.1 | - | 6 - 7 | 721 | 88 | ||
BB617492.1 | - | 4 - 5 | 532 | 88 | ||
BC062194.1 | - | 5 - 6 | 555 | 88 | ||
BC082787.1 | - | 6 - 7 | 646 | 88 | ||
BC141404.1 | - | 17 - 18 | 2229 | 9056017 | 88 | |
CD547724.1 | - | 1 - 2 | 125 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
