Assay Details
Gene Symbol: Tnfrsf25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291010.1 | NP_001277939.1 | 4 - 5 | 623 | 68 | |
GenBank mRNA | AF329969.1 | - | 4 - 5 | 627 | 68 | |
AK040632.1 | - | 3 - 4 | 592 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
