Assay Details
Gene Symbol: Pank4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK039866.1 | - | 1 - 2 | 182 | 62 | |
AK050894.1 | - | 4 - 5 | 624 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
